siRNA Details
| Gene Symbol: Ppp2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017040.1 | NP_058736.1 | 7 | 1259 | ||
| GenBank mRNA | BC085926.1 | - | 7 | 1243 | 7109733 | |
| FQ225122.1 | - | 3 | 295 | |||
| FQ231171.1 | - | 3 | 348 | |||
| M23591.1 | - | 7 | 1259 | |||
| X14087.1 | - | 7 | 1092 | |||
| X16044.1 | - | 7 | 994 | |||

Ppp2cb
Genomic Map