siRNA Details
| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017046.1 | NP_058742.1 | 2 | 173 | ||
| GenBank mRNA | EU627517.1 | - | 1 | 75 | ||
| U37539.1 | - | 2 | 127 | |||
| U37540.1 | - | 2 | 127 | |||
| X77933.1 | - | 2 | 173 | |||
| X78034.1 | - | 2 | 156 | |||

Scnn1g
Genomic Map