siRNA Details
| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017051.2 | NP_058747.1 | 2 | 256 | ||
| GenBank mRNA | BC070913.1 | - | 2 | 256 | 7104698 | |
| GQ404503.1 | - | 2 | 173 | |||
| Y00497.1 | - | 2 | 184 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sod2| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017051.2 | NP_058747.1 | 2 | 256 | ||
| GenBank mRNA | BC070913.1 | - | 2 | 256 | 7104698 | |
| GQ404503.1 | - | 2 | 173 | |||
| Y00497.1 | - | 2 | 184 | |||