siRNA Details
Gene Symbol: Tacr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017053.1 | NP_058749.1 | 2 | 790 | ||
GenBank mRNA | J05189.1 | - | 2 | 790 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
