siRNA Details
| Gene Symbol: Tacr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017053.1 | NP_058749.1 | 5 | 2075 | ||
| GenBank mRNA | J05189.1 | - | 5 | 2075 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tacr3| Gene Symbol: Tacr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017053.1 | NP_058749.1 | 5 | 2075 | ||
| GenBank mRNA | J05189.1 | - | 5 | 2075 | ||
Set Membership: |
|