siRNA Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017061.2 | NP_058757.1 | 7 | 1419 | ||
| XM_006254713.2 | XP_006254775.1 | 5 | 1502 | |||
| XM_006254714.2 | XP_006254776.1 | 6 | 1402 | |||
| XM_006254715.2 | XP_006254777.1 | 5 | 1503 | |||
| GenBank mRNA | BC078861.1 | - | 7 | 1419 | 7113042 | |
| FQ231737.1 | - | 4 | 443 | |||
| U11038.1 | - | 5 | 1387 | |||

Lox
Genomic Map