siRNA Details
| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017063.1 | NP_058759.1 | 3 | 278 | ||
| GenBank mRNA | L38644.1 | - | 3 | 278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kpnb1| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017063.1 | NP_058759.1 | 3 | 278 | ||
| GenBank mRNA | L38644.1 | - | 3 | 278 | ||