siRNA Details
| Gene Symbol: Ptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017066.2 | NP_058762.1 | 4, 5 | 519 | ||
| XM_006236290.2 | XP_006236352.1 | 4, 5 | 1903 | |||
| XM_006236294.2 | XP_006236356.1 | 4, 5 | 1499 | |||
| GenBank mRNA | BC062013.1 | - | 4, 5 | 519 | 5621277 | |
| FQ213442.1 | - | 4, 5 | 523 | |||
| FQ223195.1 | - | 4, 5 | 671 | |||
| M55601.1 | - | 4, 5 | 720 | |||

Ptn
Genomic Map