siRNA Details
| Gene Symbol: Chm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017067.1 | NP_058763.1 | 8 | 1000 | ||
| GenBank mRNA | L13722.1 | - | 8 | 1000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chm| Gene Symbol: Chm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017067.1 | NP_058763.1 | 8 | 1000 | ||
| GenBank mRNA | L13722.1 | - | 8 | 1000 | ||