siRNA Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017069.3 | NP_058765.3 | 6, 7 | 877 | ||
| XM_006229511.2 | XP_006229573.1 | 6, 7 | 989 | |||
| GenBank mRNA | X51991.1 | - | 6, 7 | 884 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra3| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017069.3 | NP_058765.3 | 6, 7 | 877 | ||
| XM_006229511.2 | XP_006229573.1 | 6, 7 | 989 | |||
| GenBank mRNA | X51991.1 | - | 6, 7 | 884 | ||