siRNA Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130710.1 | NP_001124182.1 | 5 | 479 | ||
| NM_001139499.1 | NP_001132971.1 | 5 | 718 | |||
| NM_012322.2 | NP_036454.1 | 5 | 542 | |||
| NR_024466.1 | 5 | 1289 | ||||
| GenBank mRNA | AF182291.1 | - | 5 | 503 | ||
| AJ238097.1 | - | 5 | 497 | |||
| AK024217.1 | - | 5 | 525 | |||
| BF700164.1 | - | 5 | 477 | |||

LSM5
Genomic Map