siRNA Details
| Gene Symbol: Hsd11b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017081.2 | NP_058777.1 | 5 | 1482 | ||
| GenBank mRNA | U22424.1 | - | 5 | 1483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd11b2| Gene Symbol: Hsd11b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017081.2 | NP_058777.1 | 5 | 1482 | ||
| GenBank mRNA | U22424.1 | - | 5 | 1483 | ||