siRNA Details
| Gene Symbol: Umod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017082.1 | NP_058778.1 | 10 | 2114 | ||
| XM_006230107.2 | XP_006230169.1 | 11 | 2236 | |||
| GenBank mRNA | BC081814.1 | - | 10 | 2120 | 7109450 | |
| M63510.1 | - | 10 | 2114 | |||
| S75960.1 | - | 10 | 2105 | |||

Umod
Genomic Map