siRNA Details
| Gene Symbol: Myo5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017083.1 | NP_058779.1 | 9 | 1143 | ||
| XM_006254908.2 | XP_006254970.1 | 5 | 596 | |||
| XM_008772085.1 | XP_008770307.1 | 9 | 1106 | |||
| XM_008772086.1 | XP_008770308.1 | 9 | 1106 | |||
| GenBank mRNA | U60416.1 | - | 9 | 1143 | ||

Myo5b
Genomic Map