siRNA Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017085.2 | NP_058781.2 | 4 | 949 | ||
| GenBank mRNA | EF091858.1 | - | 3 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp19a1| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017085.2 | NP_058781.2 | 4 | 949 | ||
| GenBank mRNA | EF091858.1 | - | 3 | 236 | ||