siRNA Details
| Gene Symbol: Efnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017089.2 | NP_058785.2 | 5 | 2132 | ||
| GenBank mRNA | BC061744.1 | - | 5 | 2132 | 5599058 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efnb1| Gene Symbol: Efnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017089.2 | NP_058785.2 | 5 | 2132 | ||
| GenBank mRNA | BC061744.1 | - | 5 | 2132 | 5599058 | |