siRNA Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232518.2 | XP_006232580.1 | 10 | 3869 | ||
| XM_008761107.1 | XP_008759329.1 | 8 | 3900 | |||
| XM_008761108.1 | XP_008759330.1 | 9 | 3643 | |||
| GenBank mRNA | FQ224100.1 | - | 1 | 258 | ||
| U60835.1 | - | 9 | 3597 | |||

Gucy1a3
Genomic Map