siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017094.1 | NP_058790.1 | 3 | 277 | ||
| XM_008760752.1 | XP_008758974.1 | 3 | 331 | |||
| XM_008760753.1 | XP_008758975.1 | 3 | 212 | |||
| XM_008760754.1 | XP_008758976.1 | 3 | 555 | |||
| GenBank mRNA | BC086591.1 | - | 3 | 260 | ||
| BC103659.1 | - | 3 | 239 | |||
| BC127501.1 | - | 3 | 318 | |||
| BC161852.1 | - | 3 | 264 | |||
| FQ219220.1 | - | 3 | 309 | |||
| J04811.1 | - | 3 | 277 | |||
| JF330099.1 | - | 1 | 17 | |||
| JF412704.1 | - | 2 | 87 | |||
| S49003.1 | - | 3 | 277 | |||

Ghr
Genomic Map