siRNA Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017196.3 | NP_058892.1 | 6 | 521 | ||
| XM_006256061.2 | XP_006256123.1 | 5 | 895 | |||
| XM_006256062.2 | XP_006256124.1 | 6 | 872 | |||
| XM_006256063.2 | XP_006256125.1 | 6 | 835 | |||
| XM_006256065.2 | XP_006256127.1 | 8 | 671 | |||
| GenBank mRNA | AB000818.1 | - | 6 | 519 | ||
| AF299328.1 | - | 3 | 261 | |||
| BC081822.1 | - | 4 | 1368 | |||
| D82069.1 | - | 8 | 676 | |||
| FM053942.1 | - | 6 | 508 | |||
| FQ231499.1 | - | 6 | 519 | |||
| U10894.1 | - | 3 | 275 | |||
| U17919.1 | - | 6 | 493 | |||
| U33471.1 | - | 4 | 1544 | |||

Aif1
Genomic Map