siRNA Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017196.3 | NP_058892.1 | 6 | 627 | ||
| XM_006256061.2 | XP_006256123.1 | 5 | 1001 | |||
| XM_006256062.2 | XP_006256124.1 | 6 | 978 | |||
| XM_006256063.2 | XP_006256125.1 | 6 | 941 | |||
| XM_006256065.2 | XP_006256127.1 | 8 | 777 | |||
| GenBank mRNA | AB000818.1 | - | 6 | 625 | ||
| BC081822.1 | - | 4 | 1474 | |||
| D82069.1 | - | 8 | 782 | |||
| FQ231499.1 | - | 6 | 625 | |||
| U17919.1 | - | 6 | 599 | |||

Aif1
Genomic Map