siRNA Details
Gene Symbol: Lipf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017341.2 | NP_059037.1 | 4 | 301 | ||
GenBank mRNA | A01157.1 | - | 3 | 260 | ||
X02309.1 | - | 3 | 302 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lipf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017341.2 | NP_059037.1 | 4 | 301 | ||
GenBank mRNA | A01157.1 | - | 3 | 260 | ||
X02309.1 | - | 3 | 302 |