siRNA Details
| Gene Symbol: Cdon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017358.1 | NP_059054.1 | 4 | 586 | ||
| GenBank mRNA | AF004840.1 | - | 4 | 586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdon| Gene Symbol: Cdon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017358.1 | NP_059054.1 | 4 | 586 | ||
| GenBank mRNA | AF004840.1 | - | 4 | 586 | ||