siRNA Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110782.2 | NP_001104252.1 | 1 | 352 | ||
| NM_019133.2 | NP_062006.1 | 1 | 352 | |||
| GenBank mRNA | FQ084192.1 | - | 1 | 469 | ||
| M27812.1 | - | 1 | 334 | |||
| M27924.1 | - | 1 | 334 | |||
| X04655.1 | - | 1 | 221 | |||

Syn1
Genomic Map