siRNA Details
| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019154.1 | NP_062027.1 | 2, 3 | 118 | ||
| GenBank mRNA | U67130.1 | - | 2, 3 | 118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amelx| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019154.1 | NP_062027.1 | 2, 3 | 118 | ||
| GenBank mRNA | U67130.1 | - | 2, 3 | 118 | ||