siRNA Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019378.1 | NP_062251.1 | 470 | |||
| GenBank mRNA | AF156981.1 | - | 470 | |||
| AF156982.1 | - | 470 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srcin1| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019378.1 | NP_062251.1 | 470 | |||
| GenBank mRNA | AF156981.1 | - | 470 | |||
| AF156982.1 | - | 470 | ||||