siRNA Details
| Gene Symbol: CFAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012337.2 | NP_036469.2 | 7, 8 | 911 | ||
| GenBank mRNA | AF094758.1 | - | 7, 8 | 944 | ||
| AK225566.1 | - | 7, 8 | 957 | |||
| AK225582.1 | - | 7, 8 | 957 | |||
| AK292249.1 | - | 7, 8 | 960 | |||
| AK308262.1 | - | 7, 8 | 910 | |||
| BC027874.1 | - | 7, 8 | 911 | |||
| BC089391.1 | - | 7, 8 | 1106 | 30387473 | ||
| BC109055.1 | - | 6, 7 | 886 | |||

CFAP45
Genomic Map