siRNA Details
| Gene Symbol: Gip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019630.3 | NP_062604.1 | 6 | 579 | ||
| XM_006247177.2 | XP_006247239.1 | 5 | 801 | |||
| XM_008767947.1 | XP_008766169.1 | 6 | 621 | |||
| GenBank mRNA | D11439.1 | - | 5 | 551 | ||
| L08831.1 | - | 6 | 579 | |||
| M92916.1 | - | 5 | 2751 | |||
| X66724.1 | - | 6 | 640 | |||
| Z19564.1 | - | 6 | 579 | |||

Gip
Genomic Map