siRNA Details
| Gene Symbol: Fgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020071.2 | NP_064456.2 | 2 | 155 | ||
| XM_006232524.2 | XP_006232586.1 | 2 | 173 | |||
| XM_008761078.1 | XP_008759300.1 | 2 | 112 | |||
| GenBank mRNA | AY321323.1 | - | 151 | |||
| AY325147.1 | - | 151 | ||||
| AY325153.1 | - | 151 | ||||
| BC088102.1 | - | 2 | 155 | 7371665 | ||
| FQ209924.1 | - | 2 | 162 | |||
| FQ219024.1 | - | 2 | 65 | |||
| U05675.1 | - | 2 | 156 | |||

Fgb
Genomic Map