siRNA Details
| Gene Symbol: Adcy10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021684.1 | NP_067716.1 | 3 | 380 | ||
| XM_008769677.1 | XP_008767899.1 | 2 | 2427 | |||
| XM_008769678.1 | XP_008767900.1 | 2 | 2424 | |||
| XM_008769679.1 | XP_008767901.1 | 2 | 153 | |||
| XM_008769680.1 | XP_008767902.1 | 2 | 2427 | |||
| GenBank mRNA | AF081941.1 | - | 3 | 380 | ||

Adcy10
Genomic Map