siRNA Details
| Gene Symbol: TSPAN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012339.3 | NP_036471.1 | 8 | 1600 | ||
| XM_005269666.4 | XP_005269723.1 | 6 | 2060 | |||
| XM_005269667.3 | XP_005269724.1 | 6 | 1343 | |||
| XM_011539562.2 | XP_011537864.1 | 6 | 1303 | |||
| XM_011539563.2 | XP_011537865.1 | 6 | 1514 | |||
| GenBank mRNA | AF120266.1 | - | 8 | 1591 | ||
| BC003157.1 | - | 8 | 1594 | 2958221 | ||
| BC004161.2 | - | 8 | 1594 | 2958221 | ||
| BI457070.1 | - | 191 | ||||

TSPAN15
Genomic Map