siRNA Details
| Gene Symbol: Sult1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021769.1 | NP_068537.1 | 7 | 1096 | ||
| GenBank mRNA | U32372.1 | - | 7 | 1096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1d1| Gene Symbol: Sult1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021769.1 | NP_068537.1 | 7 | 1096 | ||
| GenBank mRNA | U32372.1 | - | 7 | 1096 | ||