siRNA Details
| Gene Symbol: Cxcl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033882.1 | NP_001029054.1 | 3 | 308 | ||
| NM_001033883.1 | NP_001029055.1 | 3 | 308 | |||
| NM_022177.3 | NP_071513.1 | 3 | 308 | |||
| GenBank mRNA | AF189724.1 | - | 3 | 229 | ||
| AF209976.1 | - | 3 | 259 | |||
| AF217564.1 | - | 3 | 280 | |||
| BC078737.1 | - | 3 | 307 | 7128141 | ||
| CK475409.1 | - | 3 | 308 | |||
| CV081127.1 | - | 3 | 275 | |||

Cxcl12
Genomic Map