siRNA Details
| Gene Symbol: Gpr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022216.1 | NP_071552.1 | 1 | 953 | ||
| XM_006241735.2 | XP_006241797.1 | 2 | 1039 | |||
| XM_006241737.2 | XP_006241799.1 | 2 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr20| Gene Symbol: Gpr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022216.1 | NP_071552.1 | 1 | 953 | ||
| XM_006241735.2 | XP_006241797.1 | 2 | 1039 | |||
| XM_006241737.2 | XP_006241799.1 | 2 | 1015 | |||