siRNA Details
| Gene Symbol: Htr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022225.1 | NP_071561.1 | 1 | 104 | ||
| GenBank mRNA | X62944.1 | - | 1 | 140 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr1b| Gene Symbol: Htr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022225.1 | NP_071561.1 | 1 | 104 | ||
| GenBank mRNA | X62944.1 | - | 1 | 140 | ||
Set Membership: |
|