siRNA Details
| Gene Symbol: Stat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022380.1 | NP_071775.1 | 1425 | |||
| GenBank mRNA | X91988.1 | - | 10 | 1459 | ||
| X97541.1 | - | 1425 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stat5b| Gene Symbol: Stat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022380.1 | NP_071775.1 | 1425 | |||
| GenBank mRNA | X91988.1 | - | 10 | 1459 | ||
| X97541.1 | - | 1425 | ||||