siRNA Details
| Gene Symbol: Aldh1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022407.3 | NP_071852.2 | 9 | 972 | ||
| GenBank mRNA | AF001896.1 | - | 9 | 972 | ||
| AF001897.1 | - | 9 | 972 | |||
| AF001898.1 | - | 9 | 972 | |||
| BC061526.1 | - | 9 | 991 | 6921926 | ||
| L42009.1 | - | 9 | 968 | |||
| U79118.1 | - | 9 | 971 | |||

Aldh1a1
Genomic Map