siRNA Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022441.2 | NP_071886.1 | 8 | 1225 | ||
| XM_006242303.2 | XP_006242365.1 | 9 | 1626 | |||
| XM_006242304.2 | XP_006242366.1 | 9 | 2230 | |||
| XM_006242305.2 | XP_006242367.1 | 8 | 2245 | |||
| XM_006242307.2 | XP_006242369.1 | 8 | 2115 | |||
| XM_008765673.1 | XP_008763895.1 | 9 | 2289 | |||
| GenBank mRNA | BC083173.1 | - | 8 | 1225 | 7098096 | |
| FQ223580.1 | - | 8 | 1267 | |||
| L36088.1 | - | 9 | 1640 | |||

Acvrl1
Genomic Map