siRNA Details
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270983.1 | NP_001257912.1 | 3 | 515 | ||
NM_022498.2 | NP_071943.1 | 3 | 515 | |||
XM_008769212.1 | XP_008767434.1 | 4 | 402 | |||
GenBank mRNA | BC078825.1 | - | 3 | 442 | 7112668 | |
D90165.1 | - | 3 | 344 | |||
D90166.1 | - | 3 | 458 | |||
FN803249.1 | - | 3 | 402 |
