siRNA Details
| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022669.1 | NP_073160.1 | 1 | 112 | ||
| XM_006245148.2 | XP_006245210.1 | 2 | 302 | |||
| XM_008767195.1 | XP_008765417.1 | 2 | 436 | |||
| GenBank mRNA | BC061549.1 | - | 2 | 154 | 6919444 | |
| M93669.1 | - | 1 | 112 | |||
| X13618.1 | - | 1 | 112 | |||

Scg2
Genomic Map