siRNA Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022846.2 | NP_074037.1 | 6 | 700 | ||
| XM_006253906.1 | XP_006253968.1 | 6 | 760 | |||
| GenBank mRNA | BC097316.1 | - | 6 | 701 | 7442457 | |
| FQ219955.1 | - | 6 | 760 | |||
| FQ220067.1 | - | 6 | 759 | |||
| FQ220110.1 | - | 6 | 760 | |||
| FQ222092.1 | - | 2 | 246 | |||
| FQ222955.1 | - | 6 | 760 | |||
| FQ228468.1 | - | 2 | 330 | |||
| FQ229664.1 | - | 6 | 734 | |||
| FQ229702.1 | - | 6 | 758 | |||
| FQ229765.1 | - | 6 | 756 | |||
| FQ229882.1 | - | 6 | 760 | |||
| L06441.1 | - | 6 | 702 | |||

Prl8a2
Genomic Map