siRNA Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022847.1 | NP_074038.1 | 1 | 331 | ||
| GenBank mRNA | HM037361.1 | - | 877 | |||
| HM037362.1 | - | 877 | ||||
| L16922.1 | - | 1 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgr| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022847.1 | NP_074038.1 | 1 | 331 | ||
| GenBank mRNA | HM037361.1 | - | 877 | |||
| HM037362.1 | - | 877 | ||||
| L16922.1 | - | 1 | 331 | |||