siRNA Details
| Gene Symbol: Tspy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022923.1 | NP_075212.1 | 3, 4 | 899 | ||
| XM_008773672.1 | XP_008771894.1 | 3, 4 | 906 | |||
| GenBank mRNA | AF074880.1 | - | 3, 4 | 899 | ||
| AJ001377.1 | - | 3, 4 | 310 | |||

Tspy1
Genomic Map