siRNA Details
| Gene Symbol: Wdr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023975.1 | NP_076465.1 | 1 | 125 | ||
| XM_008772171.1 | XP_008770393.1 | 2 | 332 | |||
| GenBank mRNA | AF192379.1 | - | 1 | 125 | ||
| AF305813.1 | - | 1 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr7| Gene Symbol: Wdr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023975.1 | NP_076465.1 | 1 | 125 | ||
| XM_008772171.1 | XP_008770393.1 | 2 | 332 | |||
| GenBank mRNA | AF192379.1 | - | 1 | 125 | ||
| AF305813.1 | - | 1 | 125 | |||