siRNA Details
| Gene Symbol: Gadd45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024127.2 | NP_077041.2 | 2 | 262 | ||
| GenBank mRNA | AB102787.1 | - | 2 | 112 | ||
| AB102788.1 | - | 2 | 112 | |||
| BC081795.1 | - | 2 | 262 | 7110642 | ||
| L32591.1 | - | 2 | 130 | |||

Gadd45a
Genomic Map