siRNA Details
Gene Symbol: Ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024349.3 | NP_077325.2 | 501 | |||
GenBank mRNA | AB022701.1 | - | 463 | |||
BC089797.1 | - | 501 | 7130672 | |||
D13376.1 | - | 251 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024349.3 | NP_077325.2 | 501 | |||
GenBank mRNA | AB022701.1 | - | 463 | |||
BC089797.1 | - | 501 | 7130672 | |||
D13376.1 | - | 251 |