siRNA Details
| Gene Symbol: Gpx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030826.3 | NP_110453.3 | 2 | 731 | ||
| GenBank mRNA | BC058438.1 | - | 2 | 703 | 6918849 | |
| FQ211052.1 | - | 2 | 728 | |||
| FQ227523.1 | - | 2 | 760 | |||
| M21210.1 | - | 2 | 734 | |||

Gpx1
Genomic Map