siRNA Details
| Gene Symbol: Amy2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031502.1 | NP_113690.1 | 155 | |||
| GenBank mRNA | FQ233367.1 | - | 1 | 149 | ||
| J00703.2 | - | 155 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amy2a3| Gene Symbol: Amy2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031502.1 | NP_113690.1 | 155 | |||
| GenBank mRNA | FQ233367.1 | - | 1 | 149 | ||
| J00703.2 | - | 155 | ||||