siRNA Details
| Gene Symbol: Kras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031515.3 | NP_113703.1 | 4 | 559 | ||
| XM_008763354.1 | XP_008761576.1 | 5 | 741 | |||
| XM_008763355.1 | XP_008761577.1 | 5 | 3269 | |||
| GenBank mRNA | BC126086.1 | - | 5 | 697 | 8362025 | |
| FQ230946.1 | - | 5 | 740 | |||
| U09793.1 | - | 4 | 559 | |||

Kras
Genomic Map