siRNA Details
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031534.2 | NP_113722.2 | 9 | 1164 | ||
XM_006234620.1 | XP_006234682.1 | 8 | 1506 | |||
XM_006234621.2 | XP_006234683.1 | 9 | 1635 | |||
XM_006234622.2 | XP_006234684.1 | 8 | 1584 | |||
GenBank mRNA | CO382418.1 | - | 1 | 32 | ||
X69716.1 | - | 9 | 1164 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144783, M55512, DQ537939, BG087452.
Panther Classification:
Gene Ontology Categories:
