siRNA Details
| Gene Symbol: Scarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031541.1 | NP_113729.1 | 13 | 2344 | ||
| GenBank mRNA | AB002151.1 | - | 13 | 2344 | ||
| AY451993.1 | - | 13 | 2249 | |||
| AY682847.1 | - | 12 | 2120 | |||
| BC076504.1 | - | 13 | 2253 | 7099863 | ||
| D89655.1 | - | 13 | 2261 | |||

Scarb1
Genomic Map